Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAP155 Rabbit anti-Human, Mouse, Rat, Clone: JB40-32, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP275656
Description
SAP155 Monoclonal specifically detects SAP155 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SAP155 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:200, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50-1:200 | |
Hsh155, pre-mRNA processing 10, pre-mRNA splicing factor SF3b, 155 kDa subunit, Pre-mRNA-splicing factor SF3b 155 kDa subunit, Prp10, PRPF10, SAP 155, SAP155splicing factor 3b, subunit 1, 155kD, SF3B155, SF3b155PRP10, spliceosome associated protein 155, Spliceosome-associated protein 155, splicing factor 3B subunit 1, splicing factor 3b, subunit 1, 155kDa | |
Rabbit | |
146 kDa | |
100 μL | |
23451.0 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
JB40-32 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
SF3B1 | |
Recombinant protein within Human SAP155 aa 1-105/ 1,304. (SwissProt: O75533 Human; SwissProt: Q99NB9 Mouse) | |
Protein A purified | |
Primary | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction