Learn More
SAP30 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259195
 View more versions of this product
Catalog No. NBP259195
Add to cart
Description
Description
SAP30 Polyclonal antibody specifically detects SAP30 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation.
Specifications
Specifications
SAP30
Unconjugated
PBS with 0.05% Sodium Azide and 0.05% ProClin 300
30 kDa Sin3-associated polypeptide, histone deacetylase complex subunit SAP30, Sin3 corepressor complex subunit SAP30, Sin3A-associated protein, 30kDa, Sin3-associated polypeptide p30, sin3-associated polypeptide, 30 kDa, Sin3-associated polypeptide, 30kDa
Rabbit
IgG
50 ug
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot, ChIP assay
Western Blot 1:1000, Chromatin Immunoprecipitation 2-5 ug
Purified
SAP30
The exact immunogen is propietary information.
Protein G purified
RUO
Primary
8819