Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAP30BP Monoclonal Antibody (OTI6E10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Supplier: OriGene TA803106
Description
Induces cell death. May act as a transcriptional corepressor of a gene related to cell survival. May be involved in the regulation of beta-2-microglobulin genes.Specifications
SAP30BP | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9UHR5 | |
SAP30BP | |
Human recombit protein fragment corresponding to amino acids 1-260 of human SAP30BP produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Western Blot | |
OTI6E10 | |
Unconjugated | |
SAP30BP | |
HCNGP, HTRG, HTRP | |
Mouse | |
Affinity Chromatography | |
RUO | |
29115 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction