Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SAP30BP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584132
Description
Immunogen sequence: QELVASFSER VRNMSPDEIK IPPEPPGRCS NHLQDKIQKL YERKIKEGMD MNYIIQRKKE FRNPSIYEKL IQFCAIDELG TNYP.
Induces cell death. May act as a transcriptional corepressor of a gene related to cell survival. May be involved in the regulation of beta-2-microglobulin genes. [UniProt].
Specifications
SAP30BP | |
Polyclonal | |
Unconjugated | |
Sap30bp | |
2700016D05Rik; fb33d03; Hcngp; HSV-1 binding; HTRG; HTRP; RGD1565537; SAP30 binding protein; SAP30-binding protein; Sap30bp; transcriptional regulator protein; transcriptional regulator protein (Hcngp); transcriptional regulator protein HCNGP; Unknown (protein for MGC:142452); wu:fb33d03; zgc:55340 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29115 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UHR5 | |
Sap30bp | |
Recombinant protein corresponding to Human SAP30BP. Recombinant protein control fragment (Product #RP-103142). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction