Learn More
Greiner Bio-One Sapphire Bulk Non-Sterile Pipette Tips
Reliable fit for liquid transfers.
Specifications
Sterility Non-sterile
Material PP (Polypropylene)
Graduated Yes
Autoclavable Autoclavable
View More Specs
Products 3
Catalog Number Mfr. No. Color Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Color Volume (Metric) Price Quantity & Availability  
07-000-810
View Documents Promotion Details
Greiner Bio-One
737290
Yellow 200 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
07-000-828
View Documents Promotion Details
Greiner Bio-One
750250
Natural 1250 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
07-000-859
View Documents Promotion Details
Greiner Bio-One
771250
Natural 10 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Better recovery for small sample volumes
  • Non-Pyrogenic
  • Free of detectable DNase, RNase, human DNA
  • Tips and racks are autoclavable
  • >
Specifications
Specifications
Non-sterile
PP (Polypropylene)
Yes
Autoclavable