Learn More
Greiner Bio-One Sapphire Racked Non-Sterile Pipette Tips
Reliable fit for liquid transfers.
$59.80 - $62.80
Specifications
Sterility Non-sterile
Material PP (Polypropylene)
Graduated Yes
Color Natural
Autoclavable Autoclavable
View More Specs
Products 3
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
07-000-812
View Documents
Greiner Bio-One
738254
300 μL Case of 960 for $59.80
Add to cart
07-000-829
View Documents
Greiner Bio-One
750254
1250 μL Case of 768 for $62.80
Add to cart
07-000-861
View Documents
Greiner Bio-One
771254
10 μL Case of 960 for $59.80
Add to cart
Description
Description
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Autoclavable
  • User-friendly
  • Extended 10µL tip for better recovery of small sample volumes
Specifications
Specifications
Non-sterile
Yes
Autoclavable
PP (Polypropylene)
Natural