Learn More
Greiner Bio-One Sapphire Racked Sterile Filter Tips
Used for high recovery rate and maximum precision.
Specifications
Sterility Sterile
Filtered Filtered
Graduated Yes
Color Natural
View More Specs
Products 3
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
07-000-864
View Documents Promotion Details
Greiner Bio-One
771261
10 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
07-000-869
View Documents Promotion Details
Greiner Bio-One
773261
20 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
07-000-351
View Documents Promotion Details
Greiner Bio-One
750261
1250 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Better recovery for small sample volumes
  • Non-Pyrogenic
  • Free of detectable DNase, RNase, human DNA
  • Tips and racks are autoclavable
  • User-friendly and stackable rack
Specifications
Specifications
Sterile
Filtered
Yes
Natural