Learn More
Greiner Bio-One Sapphire Racked Sterile Low Retention Filter Tips
Used for high recovery rate and maximum precision.
Supplier: Greiner Bio-One 750265
Description
- Better recovery for small sample volumes
- Non-Pyrogenic
- Free of detectable DNase, RNase, human DNA
- Tips and racks are autoclavable
- User-friendly and stackable rack
Specifications
1250 μL | |
Sterile | |
Transparent, Insert: Blue | |
DNase, RNase, DNA, and Pyrogen Free | |
Yes | |
Polypropylene |
Filtered | |
Natural | |
Autoclavable | |
Low Retention | |
Sapphire |