Learn More
Greiner Bio-One Sapphire Racked Sterile Low Retention Filter Tips
Used for high recovery rate and maximum precision.
Specifications
Sterility Sterile
Material PP (Polypropylene)
Graduated Yes
Diameter (Metric) Outer 6.4mm
For Use With (Application) Liquid handling
View More Specs
Products 5
Catalog Number Mfr. No. Volume (Metric) Tip Style Rack Color Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Tip Style Rack Color Price Quantity & Availability  
07-000-854
View Documents
Greiner Bio-One
771235
10 μL Low Retention Transparent, Insert: Red This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
07-000-865
View Documents
Greiner Bio-One
771265
10 μL Low Retention, Extended Length Transparent, Insert: Red This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
07-000-867
View Documents
Greiner Bio-One
772265
100 μL Low Retention Transparent, Insert: Red This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
07-000-870
View Documents
Greiner Bio-One
773265
20 μL Low Retention Transparent, Insert: Red This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
07-000-350
View Documents
Greiner Bio-One
750265
1250 μL Low Retention Transparent, Insert: Blue This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Better recovery for small sample volumes
  • Non-Pyrogenic
  • Free of detectable DNase, RNase, human DNA
  • Tips and racks are autoclavable
  • User-friendly and stackable rack
Specifications
Specifications
Sterile
Yes
Liquid handling
Natural
PP (Polypropylene)
6.4mm
45.7mm
Autoclavable