Learn More
Greiner Bio-One Sapphire Refill Non-sterile Pipette Tips
Reliable fit for liquid transfers.
$67.30
Specifications
Material PP (Polypropylene)
Graduated Yes
Color Natural
Autoclavable Autoclavable
View More Specs
Products 5
Catalog Number Mfr. No. Volume (Metric) Tip Style Rack Color Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Tip Style Rack Color Price Quantity & Availability  
07-000-349
View Documents
Greiner Bio-One
771257
10 μL Extended Length Transparent, Insert: Red
Case of 960 for $67.30
Add to cart
07-000-358
View Documents
Greiner Bio-One
737257
200 μL Standard Length Transparent, Insert: Green This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
07-000-814
View Documents
Greiner Bio-One
738257
300 μL Standard Length Transparent, Insert: Green This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
07-000-857
View Documents
Greiner Bio-One
771247
10 μL Standard Length Transparent, Insert: Green This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
07-000-352
View Documents
Greiner Bio-One
750257
1250 μL Standard Length Transparent, Insert: Blue This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Autoclavable
  • User-friendly
Specifications
Specifications
PP (Polypropylene)
Yes
Natural
Autoclavable