Learn More
SAPS1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 178191AP
Catalog No. 50-173-1815
Add to cart
Description
Description
Protein phosphatase regulatory subunits, such as SAPS1, modulate the activity of protein phosphatase catalytic subunits by restricting substrate specificity, recruiting substrates, and determining the intracellular localization of the holoenzyme. SAPS1 is a regulatory subunit for the protein phosphatase-6 catalytic subunit.
Specifications
Specifications
SAPS1
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q7TSI3, Q9UPN7
KIAA1115, PP6R1, PPP6R1, SAP190, SAPS domain family member 1, SAPS domain family, member 1, SAPS1
Rabbit
IgG
150 μL
-20°C
Primary
22870, 243819
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
PPP6R1
Liquid
PPP6R1
SAPS1 Fusion Protein Ag12149
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse