Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAR1 Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | SAR1 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB028646
|
Novus Biologicals
NBP293382100UL |
100 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB028647
|
Novus Biologicals
NBP29338220UL |
20 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
SAR1 Polyclonal antibody specifically detects SAR1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
SAR1 | |
Polyclonal | |
Purified | |
RUO | |
Human, Rat | |
COPII-associated small GTPase, EC 3.6.5, masra2, SAR1 gene homolog A, SAR1 homolog A (S. cerevisiae), SAR1a gene homolog (S. cerevisiae) 1, SAR1a gene homolog 1, SAR1a gene homolog 1 (S. cerevisiae), SAR1SARA1GTP-binding protein SAR1a, Sara | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-198 of human SAR1A (NP_064535.1). | |
Primary | |
Store at -20°:C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Membrane Trafficking and Chaperones, Signal Transduction | |
PBS (pH 7.3), 50% glycerol | |
56681 | |
IgG | |
Affinity purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title