Learn More
SAR1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 153501AP
Catalog No. 50-173-0168
Add to cart
Description
Description
This gene encodes a member of the IQGAP family. The protein contains four IQ domains, one calponin homology domain, one Ras-GAP domain and one WW domain. It interacts with components of the cytoskeleton, with cell adhesion molecules, and with several signaling molecules to regulate cell morphology and motility. Expression of the protein is upregulated by gene amplification in two gastric cancer cell lines.
Specifications
Specifications
SAR1A
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P36536, Q9NR31
COPII associated small GTPase, GTP binding protein SAR1a, masra2, SAR1, SAR1 homolog A (S. cerevisiae), SAR1A, Sara, SARA1
Rabbit
IgG
150 μL
-20°C
Primary
20224, 361842, 56681
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SAR1A
Liquid
Sar1a
SAR1A Fusion Protein Ag7596
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat