Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sarcosinamide hydrochloride, 97%

Click to view available options
Quantity:
250mg
1g
5g
5325-64-4
C3H9ClN2O
124.568
MFCD00058283
VVIXOTCTYAILNP-UHFFFAOYSA-N
2-methylamino acetamide hydrochloride,h-sar-nh2.hcl,sarcosine amide hydrochloride,sarcosinamide hydrochloride,sar-nh2 hcl,2-methylamino-acetamide hcl,acetamide, 2-methylamino-, monohydrochloride,n~2~-methylglycinamide,2-methylamino acetamide hcl,2-methylamino-acetamide hydrochloride
13892956
2-(methylamino)acetamide;hydrochloride
CNCC(=O)N.Cl
5325-64-4
C3H9ClN2O
124.568
MFCD00058283
VVIXOTCTYAILNP-UHFFFAOYSA-N
2-methylamino acetamide hydrochloride,h-sar-nh2.hcl,sarcosine amide hydrochloride,sarcosinamide hydrochloride,sar-nh2 hcl,2-methylamino-acetamide hcl,acetamide, 2-methylamino-, monohydrochloride,n~2~-methylglycinamide,2-methylamino acetamide hcl,2-methylamino-acetamide hydrochloride
13892956
2-(methylamino)acetamide;hydrochloride
CNCC(=O)N.Cl
Color White to Yellow
Quantity 250mg
Linear Formula CH3NHCH2CONH2·HCl
Beilstein 3678388
Formula Weight 124.57
Physical Form Solid
Percent Purity 97%
Chemical Name or Material Sarcosinamide hydrochloride

storageNote1 : -30° to -10°C