Alfa Aesar™ Sarcosine anhydride, 99%

Manufacturer: Alfa Aesar™ A1019114

Quantity 25g

 View more versions of this product

Catalog No. AAA1019114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5076-82-4
    Molecular Formula C6H10N2O2
    Formula Weight 142.16
    MDL Number MFCD00006519
    Beilstein 123513
    Synonym 1,4-Dimethylpiperazine-2,5-dione
    Chemical Name or Material Sarcosine anhydride

    Specifications

    Formula Weight 142.16
    Molecular Formula C6H10N2O2
    Melting Point 142° to 147°C
    Beilstein 123513
    Synonym 1,4-Dimethylpiperazine-2,5-dione
    Quantity 25g
    MDL Number MFCD00006519
    CAS 5076-82-4
    Chemical Name or Material Sarcosine anhydride
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit