Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sarcosine hydrochloride, 99%

Click to view available options
Quantity:
25g
100g
637-96-7
C3H8ClNO2
125.552
MFCD00012609
WVKIFIROCHIWAY-UHFFFAOYSA-N
sarcosine hydrochloride,2-methylamino acetic acid hydrochloride,glycine, n-methyl-, hydrochloride,n-methylglycine hydrochloride,unii-w50v8r1ze9,ccris 3353,glycine, n-methyl-, hydrochloride 1:1,sarcosine, chloride,acmc-20ajra,n-methylglycine hcl
69483
2-(methylamino)acetic acid;hydrochloride
CNCC(=O)O.Cl
637-96-7
C3H8ClNO2
125.552
MFCD00012609
WVKIFIROCHIWAY-UHFFFAOYSA-N
sarcosine hydrochloride,2-methylamino acetic acid hydrochloride,glycine, n-methyl-, hydrochloride,n-methylglycine hydrochloride,unii-w50v8r1ze9,ccris 3353,glycine, n-methyl-, hydrochloride 1:1,sarcosine, chloride,acmc-20ajra,n-methylglycine hcl
69483
2-(methylamino)acetic acid;hydrochloride
CNCC(=O)O.Cl
Applications
Sarcosine hydrochloride is used in organic synthesis.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Quantity 25g
Melting Point ~173°C (decomposition)
Merck Index 14,8373
Formula Weight 125.56
Percent Purity 99%
Chemical Name or Material Sarcosine hydrochloride