Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sarcosine hydrochloride, 99%

Manufacturer:  Alfa Aesar™ B2553622

Quantity 100g

 View more versions of this product

Catalog No. AAB2553622


  Check Availability
Add to cart

Specifications

Description

Applications
Sarcosine hydrochloride is used in organic synthesis.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Description & Specifications

Chemical Identifiers

637-96-7
125.56
14,8373
Sarcosine hydrochloride
CH3NHCH2CO2H·HCl
MFCD00012609
Methylaminoacetic acid hydrochloride; N-Methylglycine hydrochloride

Specifications

637-96-7
125.56
14,8373
≈173°C (dec.)
Sarcosine hydrochloride
CH3NHCH2CO2H·HCl
MFCD00012609
Methylaminoacetic acid hydrochloride; N-Methylglycine hydrochloride
100g
99%

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit