Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sarcosine, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
107-97-1
C3H7NO2
89.094
FSYKKLYZXJSNPZ-UHFFFAOYSA-N
sarcosine,n-methylglycine,sarcosinic acid,methylglycine,glycine, n-methyl,methylaminoacetic acid,n-methylaminoacetic acid,sarcosin,2-methylamino acetic acid,methylamino acetic acid
1088
CHEBI:15611
2-(methylamino)acetic acid
CNCC(=O)O
This item is not returnable. View return policy
107-97-1
C3H7NO2
89.094
FSYKKLYZXJSNPZ-UHFFFAOYSA-N
sarcosine,n-methylglycine,sarcosinic acid,methylglycine,glycine, n-methyl,methylaminoacetic acid,n-methylaminoacetic acid,sarcosin,2-methylamino acetic acid,methylamino acetic acid
1088
CHEBI:15611
2-(methylamino)acetic acid
CNCC(=O)O

GlyT1 inhibitor

Endogenous inhibitor of GlyT1 that displays antipsychotic activity. Potentiates the action of glycine on the NMDA glycine binding site.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Formula Weight 89.09
Percent Purity >98%
Quantity 50mg
Chemical Name or Material Sarcosine

Recommended Storage : Store at Room Temperature