Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARP Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | SARP |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP170701
|
Novus Biologicals
NBP170701 |
100 μL |
Each for $436.00
|
|
NBP1707020
|
Novus Biologicals
NBP17070120UL |
20 μL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
SARP Polyclonal specifically detects SARP in Human samples. It is validated for Western Blot.Specifications
SARP | |
Polyclonal | |
Rabbit | |
ankyrin repeat domain 42 | |
ANKRD42 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
338699 | |
Synthetic peptides corresponding to ANKRD42 (ankyrin repeat domain 42) The peptide sequence was selected from the N terminal of ANKRD42)(50ug). Peptide sequence MPGVANSGPSTSSRETANPCSRKKVHFGSIHDAVRAGDVKQLSEIVCLHW. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title