Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 N Protein Monoclonal Antibody (OTI1B2), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF814453


Description
SARS-CoV-2 N Protein Monoclonal antibody specifically detects SARS-CoV-2 N Protein in Virus samples. It is validated for ELISASpecifications
SARS-CoV-2 N Protein | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
COVID19 Nucleocapsid; COVID-19 nucleocapsid; COVID19 Spike nucleocapsid; SARS-CoV-2 N | |
Full length recombinant SARS-Cov-2 N protein produced in E.coli. | |
100 μg | |
Primary | |
Virus | |
Lyophilized |
ELISA | |
OTI1B2 | |
Unconjugated | |
N | |
Mouse | |
Protein A/G | |
RUO | |
43740575 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction