Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 nsp1 Rabbit anti-SARS-CoV-2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP315978100UL
This item is not returnable.
View return policy
Description
SARS-CoV-2 nsp1 Polyclonal antibody specifically detects SARS-CoV-2 nsp1 in SARS-CoV-2 samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
SARS-CoV-2 nsp1 | |
Polyclonal | |
PBS, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
43740578 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Non-structural protein 1, nsp1, ORF1a polyprotein, ORF1ab polyprotein, pp1a, Replicase polyprotein 1a | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of coronavirus NSP1 (YP_009742608.1). | |
100 μg | |
Primary | |
SARS-CoV-2 | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction