Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 nsp12 Rabbit anti-SARS-CoV-2, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP313456
This item is not returnable.
View return policy
Description
SARS-CoV-2 nsp12 Polyclonal antibody specifically detects SARS-CoV-2 nsp12 in SARS-CoV-2 samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
SARS-CoV-2 nsp12 | |
Polyclonal | |
Western Blot 1:1000-1:10000, Immunofluorescence | |
Non-structural protein 12, nsp12, ORF1a polyprotein, ORF1ab polyprotein, pp1a, Replicase polyprotein 1a | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of SARS-CoV-2 (COVID-19) nsp12 (Strain:Wuhan-Hu-1). The exact sequence is proprietary. | |
100 μg | |
Infections (Virus Bacteria and Parasites) | |
43740578 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunofluorescence | |
Unconjugated | |
PBS, 1% BSA, 20% Glycerol (pH7) | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
SARS-CoV-2 | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title