Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 nsp7 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP307048V
This item is not returnable.
View return policy
Description
SARS-CoV-2 nsp7 Polyclonal antibody specifically detects SARS-CoV-2 nsp7 in SARS-CoV-2 samples. It is validated for ELISA, Immunohistochemistry, ImmunofluorescenceSpecifications
SARS-CoV-2 nsp7 | |
Polyclonal | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Rabbit | |
Peptide Affinity-purified | |
RUO | |
Primary | |
SARS-CoV-2 | |
Purified |
ELISA, Immunohistochemistry, Immunofluorescence | |
DyLight 405 | |
Non-structural protein 7, nsp7 replicase | |
This antibody was raised against a peptide corresponding to 12 amino acids near the carboxyl terminus of the SARS-CoV-2 (COVID-19) NSP7 protein. The immunogen is located within the last 50 amino acids of the SARS-CoV-2 (COVID-19) NSP7 protein. | |
0.1 mL | |
Infections (Virus Bacteria and Parasites) | |
43740578 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction