Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 nsp8 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP307046MFV450
Description
SARS-CoV-2 nsp8 Polyclonal antibody specifically detects SARS-CoV-2 nsp8 in SARS-CoV-2 samples. It is validated for ELISA, ImmunohistochemistrySpecifications
| SARS-CoV-2 nsp8 | |
| Polyclonal | |
| ELISA, Immunohistochemistry | |
| Non-structural protein 8, nsp8 replicase | |
| This antibody was raised against a peptide corresponding to 14 amino acids near the center of SARS-CoV-2 (COVID-19) NSP8 protein. The immunogen is located within 60-110 amino acids of SARS-CoV-2 (COVID-19) NSP8. | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| 43740578 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA, Immunohistochemistry | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| SARS-CoV-2 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction