Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 Nucleocapsid Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300510AF750
This item is not returnable.
View return policy
Description
SARS-CoV-2 Nucleocapsid Polyclonal antibody specifically detects SARS-CoV-2 Nucleocapsid in SARS-CoV-2, Virus samples. It is validated for Western BlotSpecifications
SARS-CoV-2 Nucleocapsid | |
Polyclonal | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Rabbit | |
Immunogen Affinity-purified | |
RUO | |
Primary | |
SARS-CoV-2, Virus | |
Purified |
Western Blot | |
Alexa Fluor 750 | |
2019-nCoV N Protein, 2019-nCoV Nucleocapsid Protein, COVID-19 N protein, COVID-19 nucleocapsid protein, Human coronavirus Nucleoprotein, nucleocapsid phosphoprotein, Nucleocapsid protein, Nucleoprotein, ORF9; structural protein, SARS-CoV-2, SARS-CoV-2 Nucleocapsid protein, SARS-CoV-2 N protein, SARSCoV2 Nucleoprotein, Severe Acute Respiratory Syndrome Coronavirus 2 Nucleocapsid Protein | |
Full length recombinant SARS-CoV-2 nucleocapsid protein. | |
0.1 mL | |
Virology Bacteria and Parasites | |
43740575 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction