Learn More
ACROBiosystems ACROBiosystems SARS-CoV-2 Nucleocapsid protein (D63G, R203M, D377Y, R385K), His Tag
Supplier:  ACROBiosystems NUNC52HP100UG
Encompass_Preferred
Recombinant Protein;100UG;SARS-CoV-2 Nucleocapsid protein, His Tag (NUN-C52Hp) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 419 (Accession # QHO62115.1). The mutations (D63G, R203M, D377Y, R385K) were identified in the SARS-CoV-2 variants which emerged in India (known as B.1.617.2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 47.2 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized SARS-CoV-2 Nucleocapsid protein, His Tag (Cat. No. NUN-C52Hp) at 1 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Nucleocapsid Antibody, Human IgG1 (Cat. No. NUN-S41A1) with a linear range of 0.2-1 ng/mL (QC tested).;Immobilized SARS-CoV-2 Nucleocapsid protein, His Tag (Cat. No. NUN-C52Hp) at 1 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Nucleocapsid Antibody, Chimeric mAb, Human IgG1 (AM223) (Cat. No. NUN-M223) with a linear range of 0.2-1 ng/mL (Routinely tested).
Catalog No. 50-210-6986