Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 Nucleocapsid Rabbit anti-SARS-CoV-2, DyLight 755, Clone: 1, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP290988IR

Catalog No. NB041462


Add to cart

Description

Description

SARS-CoV-2 Nucleocapsid Monoclonal antibody specifically detects SARS-CoV-2 Nucleocapsid in SARS-CoV-2 samples. It is validated for Western Blot, ELISA
Specifications

Specifications

SARS-CoV-2 Nucleocapsid
1
Western Blot, ELISA
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Monoclonal
ELISA, Western Blot
DyLight 755
50mM Sodium Borate
2019-nCoV N Protein, 2019-nCoV Nucleocapsid Protein, COVID-19 N protein, COVID-19 nucleocapsid protein, Human coronavirus Nucleoprotein, Nucleocapsid protein, Nucleoprotein, SARS-CoV-2 Nucleocapsid protein, SARS-CoV-2 N protein, SARSCoV2 Nucleoprotein, Severe Acute Respiratory Syndrome Coronavirus 2 Nucleocapsid Protein
Recombinant SARS-CoV Nucleoprotein/ NP Protein
Protein A purified
RUO
Primary
SARS-CoV-2
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit