Learn More
Microbiologics™ SARS-CoV-2 Process Control Swab
SARS-CoV-2 Process Control can be used to monitor the extraction, amplification and detection of SARS-CoV-2 molecular applications. Routine use of external quality controls monitor day to day performance of molecular testing procedures.
Manufacturer:  Microbiologics™ HE0063S
Catalog No. 23-047-381
Add to cart
Description
Description
  • Each kit contains 5 individually packaged lyophilized swabs.