Learn More
SARS-CoV-2 Spike (N501Y Mutant) Monoclonal Antibody (HL1425), Invitrogen™
Rabbit Monoclonal Antibody
Supplier:  Thermo Scientific MA547040
Encompass
Catalog No. PIMA547040
Add to Cart
Description
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
SARS-CoV-2 Spike (N501Y Mutant) Monoclonal antibody specifically detects SARS-CoV-2 Spike (N501Y Mutant) in Virus samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SARS-CoV-2 Spike (N501Y Mutant)
Monoclonal
1 mg/mL
PBS with no preservative
Rabbit
Protein A
RUO
Virus
Antibody
IgG
Western Blot
HL1425
Unconjugated
2019-nCoV Spike; COVID-19 Spike; SARS-CoV-2 Spike
Carrier-protein conjugated synthetic peptide surrounding mutant N501Y of SARS-CoV-2 (COVID-19) Spike. The exact sequence is proprietary.
100 μL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Recommendations
Recommendations