Learn More
SARS-CoV-2 Spike Protein RBD Recombinant Llama Monoclonal Antibody (A10), Invitrogen™
Llama Recombinant Monoclonal Antibody
Supplier:  Thermo Scientific MA549345
Encompass
Catalog No. PIMA549345
Add to Cart
Description
Description
SARS-CoV-2 Spike Protein RBD Recombinant Monoclonal antibody specifically detects SARS-CoV-2 Spike Protein RBD in Virus samples. It is validated for ELISA, Neutralization
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SARS-CoV-2 Spike Protein RBD
Recombinant Monoclonal
Unconjugated
Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2
SARS-CoV-2 Spike S2 protein containing C-terminal His Tag. The protein was expressed in human 293 cells (HEK293). It contains amino acids Ser 686-Pro 1213.
100 μg
Primary
-20° C, Avoid Freeze/Thaw Cycles
scFv
ELISA, Neutralization
A10
PBS with no preservative
Llama
Affinity chromatography
RUO
Virus
Liquid
Recommendations
Recommendations