Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 151621AP

Catalog No. 50-173-0031


Due to product restrictions, please to purchase or view availability for this product.


Description

Description

This gene belongs to the class II amino-acyl tRNA family. The encoded enzyme catalyzes the transfer of L-serine to tRNA (Ser) and is related to bacterial and yeast counterparts.
Specifications

Specifications

SARS
0.16 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P26638, P49591, Q6P799
SARS, Serine tRNA ligase, SERRS, SERS, seryl tRNA synthetase, Seryl tRNA(Ser/Sec) synthetase
Rabbit
IgG
150 μL
-20°C
Primary
20226, 266975, 6301
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SARS
Liquid
SARS
SARS Fusion Protein Ag7270
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit