Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sars Spike Protein Mouse anti-Human, Alexa Fluor™ 532, Clone: 17F706, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP224942AF532

Catalog No. NP224942A32


Add to cart

Description

Description

Sars Spike Protein Monoclonal antibody specifically detects Sars Spike Protein in Human samples. It is validated for Western Blot.
Specifications

Specifications

Sars Spike Protein
17F706
Western Blot
Affinity Purified, Conjugated
Mouse
IgG3 Kappa
0.1 mL
Primary
Human
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
E2, E2 glycoprotein, Human coronavirus spike glycoprotein, Peplomer protein, S, S glycoprotein, Severe acute respiratory syndrome spike glycoprotein, Severe acute respiratory syndrome virus spike glycoprotein, Spike glycoprotein, VGL2
Amino acids 1124-1140 of human SARS-Spike protein were used as the immunogen
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit