Learn More
Sartorius Sartoclear Dynamics: Lab V
For simultaneous clarification and sterile filtration of 50 mL to 1 L mammalian cell cultures in one step without centrifugation.
$182.00 - $828.00
Specifications
For Use With (Application) | mammalian cell harvesting/culture |
---|---|
Product Type | Sartoclear Dynamics&trade: Lab V kits have been designed for cell harvesting of mammalian cell cultures, such as CHO, HEK, hybridomas and many others, with high cell densities. |
Quantity | 12 x 1L Filter Units + 40g Filter Aid |
Storage Requirements | ambient |
Filter Type | Diatomaceous Earth/Heavy metal-free polystyrene/Heavy metal-free polyethylene |
Catalog Number | Mfr. No. | Bed Volume | Packaging Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Bed Volume | Packaging Quantity | Price | Quantity & Availability | ||||
14-558-715
|
Sartorius
SDLV100040CE |
1,000mL | 40g |
Pack of 12 for $489.00
|
|
14-558-716
|
Sartorius
SDLV100020CE |
1,000mL | 20g |
Pack of 12 for $450.00
|
|
14-558-717
|
Sartorius
SDLV100010C2 |
1,000mL | 10g |
Pack of 24 for $828.00
|
|
14-558-718
|
Sartorius
SDLV050020CE |
500mL | 20g |
Pack of 12 for $331.00
|
|
14-558-719
|
Sartorius
SDLV050010C2 |
500mL | 10g |
Pack of 24 for $577.00
|
|
14-558-720
|
Sartorius
SDLV050005C2 |
500mL | 5g |
Pack of 24 for $535.00
|
|
14-558-721
|
Sartorius
SDLV025010C2 |
250mL | 10g |
Pack of 24 for $470.00
|
|
14-558-722
|
Sartorius
SDLV025005C2 |
250mL | 5g |
Pack of 24 for $416.00
|
|
14-558-723
|
Sartorius
SDLV015005C2 |
150mL | 5g |
Pack of 24 for $388.00
|
|
14-558-724
|
Sartorius
SDLV015002CE |
150mL | 2g |
Pack of 12 for $182.00
|
|
Description
Sartoclear Dynamics Lab V kits are designed for simultaneous clarification and sterile filtration of 50 mL to 1 L of mammalian cell culture broth to prepare samples for subsequent downstream processing steps, such as protein purification and concentration. The convenient kits combine pouches of filter aid for clarification and vacuum filtration units for sterile filtration.
The Sartoclear Dynamics Lab V Kits combine the use of a filter aid for clarification and a vacuum filtration unit with a 0.22 µm polyethersulfone membrane for sterile filtration of up to 1 L mammalian cell cultures. The ready-to-use filter aid is supplied in dust-free and gamma-irradiated pouches.
The Sartolab RF bottletop filters with receiver flasks, sterilized by gamma irradiation are
available in volume capacities ranging from 150 mL to 1,000 mL.
- Clarify high-density mammalian cell culture in minutes
- Out-of-the-box solution
- Fast and effortless
- No need for centrifugation
Ten kits are available to meet all your needs based on your
filtration volume and the cell density of your mammalian
cell cultures.
Specifications
mammalian cell harvesting/culture | |
12 x 1L Filter Units + 40g Filter Aid | |
Diatomaceous Earth/Heavy metal-free polystyrene/Heavy metal-free polyethylene |
Sartoclear Dynamics&trade: Lab V kits have been designed for cell harvesting of mammalian cell cultures, such as CHO, HEK, hybridomas and many others, with high cell densities. | |
ambient |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.