Learn More
Sartorius™ Qualitative Grade 1290 Filter Papers
Manufacturer:  Sartorius™ FT3208150
 View more versions of this product
Catalog No. 14-555-058
Add to cart
Description
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.
Specifications
Specifications
100 per pack
1290
Disc
150mm
3 to 5μm
0.21mm