Learn More
Sartorius™ Qualitative Grade 1290 Filter Papers
Manufacturer:  Sartorius™ FT3208185
 View more versions of this product
Catalog No. 14-555-057
Add to cart
Description
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.
Specifications
Specifications
100 per pack
1290
Disc
185mm
3 to 5μm
0.21mm
Documents
Promotions
Promotions
  • Sartorius , Fisher Scientific , Fisherbrand
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.
    06-30-2021