Learn More
Sartorius Qualitative Grade 1292 Filter Papers
These qualitative filter papers are essentially used for analytical purposes and routine analyses, whenever no gravimetric analyses are required. They are wet-strengthened and can be used for pressure and vacuum filtration.
Supplier: Sartorius FT3210055
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.Specifications
Disc | |
50 sec. | |
100/Pk. | |
55 mm |
5 to 8 μm | |
1292 | |
0.17 mm |