Learn More
Sartorius™ Qualitative Grade 292 a Filter Papers
Manufacturer:  Sartorius™ FT3215240
 View more versions of this product
Catalog No. 14-555-025
Add to cart
Description
Description
  • Pure cotton linters or cotton linters with refined pulp
  • No additives, such as wet-strengthening agents
Specifications
Specifications
100 per pack
292 a
Disc
240mm
4 to 7μm
0.19mm