Learn More
Sartorius Qualitative Grade 292 a Filter Papers
These filter papers are especially suitable for qualitative analyses that require a low ash content.
Supplier: Sartorius FT3215055
Description
- Pure cotton linters or cotton linters with refined pulp
- No additives, such as wet-strengthening agents
Specifications
Disc | |
97 sec. | |
100/Pk. | |
55 mm |
4 to 7 μm | |
292 a | |
0.19 mm |