Learn More
Sartorius™ Qualitative Grade 292a Filter Papers
These filter papers are especially suitable for qualitative analyses that require a low ash content.
Manufacturer:  Sartorius™ FT4215150
Catalog No. 14-555-006
Add to cart
Description
Description
  • Pure cotton linters or cotton linters with refined pulp
  • No additives, such as wet-strengthening agents
Specifications
Specifications
100 per pack
292 a
Folded
150mm
4 to 7μm
0.19mm
Documents
Promotions
Promotions
  • Sartorius , Fisher Scientific , Fisherbrand
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.
    06-30-2021