Learn More
Sartorius™ Qualitative Grade 293 Filter Papers
Manufacturer:  Sartorius™ FT3211090
 View more versions of this product
Catalog No. 14-555-037
Add to cart
Description
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.
Specifications
Specifications
100 per pack
293
Disc
90mm
1 to 2μm
0.15mm