Learn More
Sartorius™ Qualitative Grade 34/N Filter Papers
Manufacturer:  Sartorius™ FT4478320
 View more versions of this product
Catalog No. 14-555-002
Add to cart
Description
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content between 0.1 - 0.15%. Wet-strengthened.
Specifications
Specifications
100 per pack
34/N
0.28mm
320mm
Folded
Documents
Promotions
Promotions
  • Sartorius , Fisher Scientific , Fisherbrand
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.
    06-30-2021