Learn More
Sartorius Qualitative Grade 37/N Filter Papers
Manufacturer: Sartorius FT3480150
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content between 0.1 - 0.15%. Wet-strengthened.Specifications
50 per pack | |
37/N | |
0.5mm |
150mm | |
Disc |
Promotions
-
Sartorius
,
Fisher Scientific
,
Fisherbrand
Get a Pipette Multipack with Purchase
Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.: 06-30-2021