Learn More
Sartorius™ Qualitative Grade 5 H/N Filter Papers
This paper grade is generally used for analytical and technical purposes. It has a fast flow rate due to its structure.
Manufacturer:  Sartorius™ FT4423150
Catalog No. 14-555-004
Add to cart
Description
Description
  • Made of refined pulp and cotton linters with an > 95% α-cellulose content
  • Ash content between 0.1 - 0.15%
  • Wet-strengthened
Specifications
Specifications
100 per pack
5 H/N
0.28mm
150mm
Folded
Documents
Promotions
Promotions
  • Sartorius , Fisherbrand , Fisher Scientific
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.
    06-30-2021