Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius™ Accessories for Combisart™ Systems: Dosing Syringe

Dosing syringe for use with Sartorius Combisart Systems

Manufacturer:  Sartorius™ 166852

Catalog No. 14-555-901


Add to cart

Description

Description

  • Use to moisten nutrient pads with water
  • Capacity, 0.5 - 5mL

Specifications

Specifications

0.01 to 0.17 oz.
Used to moist the nutrient pads with Water
Dosing Syringe
0.5 to 5mL
Combisart™ system
Documents
Promotions

Promotions

  • Sartorius , Fisherbrand , Fisher Scientific
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.

    06-30-2021

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit