Learn More
Sartorius arium™ 615DI Deionization Cartridges
Supplier:  Sartorius 615DI004
 View more versions of this product
Catalog No. 14-556-232
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
arium™ 615DI cartridges are designed to produce highly deionised water. The DI cartridges contain highly effective ion exchange resins to remove cations and anions from the water by exchanging H+ and OH–. The uniform and compact mixed bed ensures a reliable desalination and an optimal utilisation of the resin material, reducing conductivity by up to 0.1μS/cm.
  • Compact design, easy to install
  • Reliable procedure, simply replace the resin when its capacity is exhausted
  • Conductivity typically <0.1μS/cm
  • High efficiency
TRUSTED_SUSTAINABILITY
Specifications
Specifications
White
35°C
Set of tube connector
Plastic
40 cm
Cartridge, Deionization
40
0.75 in.
5 L
23 cm
Product Suggestions
Product Suggestions