Learn More
Sartorius™ arium™ 615DI Deionization Cartridges
Manufacturer:  Sartorius™ 615DI007
 View more versions of this product
Catalog No. 14-556-233
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
arium™ 615DI cartridges are designed to produce highly deionised water. The DI cartridges contain highly effective ion exchange resins to remove cations and anions from the water by exchanging H+ and OH–. The uniform and compact mixed bed ensures a reliable desalination and an optimal utilisation of the resin material, reducing conductivity by up to 0.1µS/cm.
  • Compact design, easy to install
  • Reliable procedure, simply replace the resin when its capacity is exhausted
  • Conductivity typically <0.1µS/cm
  • High efficiency
Specifications
Specifications
White
0.75 in.
Deionization cartridge
300
23cm
Set of tube connector
12L
35°C
140cm
Plastic
Documents
Promotions
Promotions
  • Sartorius , Fisherbrand , Fisher Scientific
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.
    06-30-2021