Learn More
Sartorius™ arium™ advance RO Type 3 Reverse Osmosis Water Systems
Provide Type 3 reverse osmosis water of the highest quality with the Sartorius™ arium™ advance RO Type 3 Reverse Osmosis Water System.
Manufacturer:  Sartorius™ H2ORO2B
 View more versions of this product
Catalog No. 14-556-228
Add to cart
Description
Description
  • Innovative iJust technology
  • Uses unique bagtank technology with dispense gun connections
  • User-friendly, easy operation
  • Unique pre-filter technology with activated carbon, phosphate-free catalyst and 5µm particle filter
Benefits:
  • Optimized water consumption
  • Meets customer's lab requirements and saves maintenance time
  • No user manual needed, intuitive operation of the menu
  • Highly efficient reduces colloids and chlorine, oxidizing agents, organic and water-soluble heavy metals
Specifications
Specifications
Type 3
16L/hour
Less than 1 CFU/1000mL
50cm
35cm
50/60Hz
2 to 35°C
Greater than 99%
17.75 in.
Less than 1 NTU
Feed water for ultrapure water system, rinsing Glassware and laboratory vessels, feed water for humidifiers, autoclaves and dish washer
80%
4 to 10
35.6 to 86°F
≤ 3L/min.
Particle filtration, spherical activated Carbon, catalyst, reverse osmosis, optional final particle and sterile filtration
48.5 lb.
Reverse osmosis water system
Less than 0.05 MO●cm for product water
Less than 2000ppb
19.7 in.
13.7 in.
100/240V
35.6 to 95°F
Greater than 99%
Greater than 99%
45cm
5 to 45°C
100/240V 50/60Hz
2 to 30°C
Less than 1/mg
Wall
Less than 20μS/cm for product water, less than 1500μS/cm for feed water
<4
22kg
Documents
Promotions
Promotions
  • Sartorius , Fisherbrand , Fisher Scientific
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.
    06-30-2021