Learn More
Sartorius arium™ Comfort I Combined System (TYPE 1 - Ultrapure Water and TYPE 3 - RO Water)
Use the Sartorius arium™ Comfort I Combined System for producing ASTM Type 1 ultrapure water and Type 3 pure water.
Supplier: Sartorius H2OI2TOCB
Description
arium comfort I systems produce ASTM Type 1 ultrapure water and Type 3 pure water. The systems features the latest reverse osmosis technology, and a unique filter cartridge for producing ultrapure water of the highest quality, delivering consistently high-grade water quality at rates of up to 120l/h. The quality of this water even exceeds the ASTM Type 1 specifications. The system's integrated UV lamp (185/254 nm) prevents microbiological growth and reduces the TOC content to a minimum. The current TOC value is continuously checked by an integrated TOC monitor specially developed for this ultrapure water system and is continuously displayed.
- Reliable TOC content of <2 ppb for reproducible results
- Automatically optimised water consumption with iJust
- Easy-to-use - unique glass display with touch function
Benefits:
- Produces Type 3 and Type ASTM 1 water from tap water all in one unit; adapted to the customer's needs; the display will conveniently remain at eye level
- Optimized water consumption
- Meet customer's lab requirements and saves maintenance time
- Safe operation
- Reliable TOC reduction
- Supports reliable recording of important data
- Easy operation
- HPLC
- GC-MS, AAS, ICP-MS
- Ion chromatography
- TOC-Analysis
- Photometry
- Buffer and pH-solutions
- Feed water for ultrapure water systems, rinsing glassware and laboratory vessels, feed water for humidifiers, autoclaves and dish washer
Specifications
0.055 Product water, Less than1500 feed water | |
Greater than99% | |
120 (ASTM type 1), 16 (Type 3) | |
Type 1 to 3 | |
<2 product water, <2000 feed water | |
35.6°F to 86°F | |
Feed water for ultrapure water system, rinsing Glassware and laboratory vessels, feed water for humidifiers, autoclaves and dish washer | |
TOC monitor | |
<1/mL | |
Adsorption by spherical activated Carbon, catalyst, reverse osmosis, deionization, UV radiation, optional final particle and sterile filtration | |
35.6°F to 95°F | |
<4 | |
17.12 in. | |
19.7 in. | |
18.74 in. | |
68.3 lb. | |
100/240 V 50/60 Hz | |
50/60 Hz |
Greater than99% | |
<1 CFU/L | |
Up to 2 L/min. (ASTM type 1), Less than 3 L/min. (Type 3) | |
18.2 Mohm.cm product water | |
Arium Comfort I Combined System | |
2°C to 30°C | |
80% | |
Wall | |
4 to 10 | |
5°C to 45°C | |
2°C to 35°C | |
<1 | |
43.5 cm | |
50.1 cm | |
47.6 cm | |
31 kg | |
100/240 V |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.