Learn More
Sartorius™ arium™ Multifunction Stand
arium™ Multifunction Stand includes a height-adjustable tripod, a slide with holder for the display/dispense unit and operating instructions.
Manufacturer:  Sartorius™ H2OPROADM1
Catalog No. 14-556-273
Add to cart
Description
Description
  • Visual quality control directly at the water-dispensing port
  • System control directly at the work place
  • Optimal customization to the variously sized sampling vessels
  • Water dispensing using the slider
  • Radius extends up to 2.5 meters
  • Sterile-grade filters connectable
Specifications
Specifications
59.5cm
25.5cm
Aluminum
arium™ Water Purification System
Connection to Arium™ Built-In system
Display mounting kit
22cm
Stand
Documents
Promotions
Promotions
  • Sartorius , Fisher Scientific , Fisherbrand
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.
    06-30-2021