Sartorius™ arium™ Pro ASTM TYPE 1 - Ultrapure Water Systems: Benchtop, Low Endotoxin

Sartorius arium™ Pro ASTM TYPE 1 - Ultrapure Water Systems lets users produce ASTM type 1 water for critical analysis like HPLC, PCR, antibody production, cell culture media preparation or electrophoresis.

Manufacturer: Sartorius™ H2OPROVFD

 View more versions of this product

Catalog No. 14-556-224

  Check Availability
Add to Cart

    Specifications

    Product Type Arium™ Pro ASTM type 1 UF ultrapure water system
    DNase <0.024
    Humidity Range 80%
    Feedwater Temperature (Metric) 2-30°C
    pH Range 4-10
    Particle Size <1/mL
    Liquid Flow Rate 1.7
    Range (Conductivity) 0.055
    Weight (English) 59.5 to 63.9 lb.
    Product Water Type Type 1
    Height (English) 19.37 in.
    Height (Metric) 49.2cm
    Width (English) 13.7 in.
    Width (Metric) 35cm
    Resistivity 18.2
    Voltage 100/240V
    Hertz 50/60Hz
    Temperature Range (English) 35.6-95°F
    Temperature Range (Metric) 2-35°C
    Depth (English) 17.75 in.
    Depth (Metric) 45.1cm
    Daily Output 120
    Turbidity Less than1
    Storage Requirements 5-45°C
    Certifications/Compliance ASTM
    For Use With (Application) Standard, cell culture and chromatographic
    Electrical Requirements 100/240V 50/60Hz
    Includes Wall mounting kit, dispenser unit
    Total Organic Carbon <2 Product water, <50 feed water
    Feedwater Temperature (English) 35.6-86°F
    Mounting Bench integrated
    RNase (Metric) <0.004
    Weight (Metric) 27 to 29kg
    Endotoxin Concentration <0.001
    Bacterial Content Less than1CFU/1000mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit